Anti-PGP Rabbit Polyclonal Antibody

Supplier: Proteintech
25081-1-AP
10155-414EA 635.16 CAD
10155-414
Anti-PGP Rabbit Polyclonal Antibody
Antibodies
The PGP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PGP. This antibody recognizes human, mouse antigen. The PGP antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: PGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR