Anti-KLHL28 Rabbit Polyclonal Antibody

Supplier: Proteintech
25224-1-AP
10155-502EA 635.16 CAD
10155-502
Anti-KLHL28 Rabbit Polyclonal Antibody
Antibodies
The KLHL28 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human KLHL28. This Antibody recognizes human, mouse Antigen. The KLHL28 Antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: KLHL28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR