Anti-KIAA0430 Rabbit Polyclonal Antibody

Supplier: Proteintech
25233-1-AP
10155-504EA 635.16 CAD
10155-504
Anti-KIAA0430 Rabbit Polyclonal Antibody
Antibodies
The KIAA0430 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human KIAA0430. This Antibody recognizes human, mouse Antigen. The KIAA0430 Antibody has been validated for the following applications: ELISA, WB, IHC, IP analysis.

WB 1 : 1000

Type: Primary
Antigen: KIAA0430
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR