Anti-BTNL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25110-1-AP
10155-432EA 818.44 CAD
10155-432
Anti-BTNL2 Rabbit Polyclonal Antibody
Antibodies
The BTNL2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BTNL2. This antibody recognizes human, mouse antigen. The BTNL2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: BTNL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR