Anti-TPTE2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25120-1-AP-150UL
10155-442EA 635.16 CAD
10155-442
Anti-TPTE2 Rabbit Polyclonal Antibody
Antibodies
The TPTE2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TPTE2. This antibody recognizes human antigen. The TPTE2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IHC 1 : 50

Type: Primary
Antigen: TPTE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR