Anti-DAPK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
25136-1-AP
10155-450EA 635.16 CAD
10155-450
Anti-DAPK1 Rabbit Polyclonal Antibody
Antibodies
The DAPK1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human DAPK1. This antibody recognizes human antigen. The DAPK1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: DAPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR