Anti-ANXA13 Rabbit Polyclonal Antibody

Supplier: Proteintech
25153-1-AP-150UL
10155-462EA 635.16 CAD
10155-462
Anti-ANXA13 Rabbit Polyclonal Antibody
Antibodies
The Annexin A13 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Annexin A13. This antibody recognizes human, mouse antigen. The Annexin A13 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

LL

Type: Primary
Antigen: ANXA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR