Anti-FAM105A Rabbit Polyclonal Antibody

Supplier: Proteintech
25668-1-AP
10155-748EA 635.16 CAD
10155-748
Anti-FAM105A Rabbit Polyclonal Antibody
Antibodies
The FAM105A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human FAM105A. This antibody recognizes human antigen. The FAM105A antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 600 ; IF 1 : 25

Type: Primary
Antigen: FAM105A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR