Anti-CDC14B Rabbit Polyclonal Antibody

Supplier: Proteintech
25187-1-AP
10156-276EA 635.16 CAD
10156-276
Anti-CDC14B Rabbit Polyclonal Antibody
Antibodies
The CDC14B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CDC14B. This antibody recognizes human antigen. The CDC14B antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: CDC14B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR