Anti-ANKRD33 Rabbit Polyclonal Antibody

Supplier: Proteintech
25202-1-AP-150UL
10155-488EA 635.16 CAD
10155-488
Anti-ANKRD33 Rabbit Polyclonal Antibody
Antibodies
The ANKRD33 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ANKRD33. This antibody recognizes human antigen. The ANKRD33 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 200 ; WB 1 : 2000

Type: Primary
Antigen: ANKRD33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR