Anti-FAM98C Rabbit Polyclonal Antibody

Supplier: Proteintech
25274-1-AP
10155-534EA 635.16 CAD
10155-534
Anti-FAM98C Rabbit Polyclonal Antibody
Antibodies
The FAM98C Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human FAM98C. This Antibody recognizes human Antigen. The FAM98C Antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: FAM98C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR