Anti-ATXN8 Rabbit Polyclonal Antibody

Supplier: Proteintech
18793-1-AP
10154-842EA 635.16 CAD
10154-842
Anti-ATXN8 Rabbit Polyclonal Antibody
Antibodies
The Twinkle antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Twinkle. This antibody recognizes human antigen. The Twinkle antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 400

Type: Primary
Antigen: ATXN8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR