Anti-CHERP Rabbit Polyclonal Antibody

Supplier: Proteintech
25389-1-AP-150UL
10155-596EA 635.16 CAD
10155-596
Anti-CHERP Rabbit Polyclonal Antibody
Antibodies
The CHERP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CHERP. This antibody recognizes human antigen. The CHERP antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600

Type: Primary
Antigen: CHERP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR