Anti-AFM Rabbit Polyclonal Antibody

Supplier: Proteintech
25280-1-AP
10156-122EA 635.16 CAD
10156-122
Anti-AFM Rabbit Polyclonal Antibody
Antibodies
The AFM Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human AFM. This Antibody recognizes human Antigen. The AFM Antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 1000

Type: Primary
Antigen: AFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR