Anti-KIAA0907 Rabbit Polyclonal Antibody

Supplier: Proteintech
25419-1-AP-150UL
10156-300EA 635.16 CAD
10156-300
Anti-KIAA0907 Rabbit Polyclonal Antibody
Antibodies
The KIAA0907 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human KIAA0907. This antibody recognizes human, mouse antigen. The KIAA0907 antibody has been validated for the following applications: ELISA, IHC, WB, IF analysis.

IF 1 : 25 ; IHC 1 : 50 ; WB 1 : 300

Type: Primary
Antigen: KIAA0907
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR