Anti-C19orf70 Rabbit Polyclonal Antibody

Supplier: Proteintech
25514-1-AP
10155-660EA 635.16 CAD
10155-660
Anti-C19orf70 Rabbit Polyclonal Antibody
Antibodies
The C19orf70 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C19orf70. This antibody recognizes human, mouse antigen. The C19orf70 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: C19orf70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR