Anti-KIAA0494 Rabbit Polyclonal Antibody

Supplier: Proteintech
19569-1-AP-150UL
10154-860EA 635.16 CAD
10154-860
Anti-KIAA0494 Rabbit Polyclonal Antibody
Antibodies
The KIAA0494 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human KIAA0494. This antibody recognizes human antigen. The KIAA0494 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 500 ; IHC 1 : 50

Type: Primary
Antigen: KIAA0494
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR