Anti-ABCB6 Rabbit Polyclonal Antibody

Supplier: Proteintech
51007-1-AP-150UL
10156-144EA 635.16 CAD
10156-144
Anti-ABCB6 Rabbit Polyclonal Antibody
Antibodies
The ABCB6 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ABCB6. This antibody recognizes human, mouse, rat antigen. The ABCB6 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600

Type: Primary
Antigen: ABCB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR