Anti-EHD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
25320-1-AP
10155-556EA 635.16 CAD
10155-556
Anti-EHD3 Rabbit Polyclonal Antibody
Antibodies
The EHD3 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human EHD3. This Antibody recognizes human, mouse Antigen. The EHD3 Antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: EHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR