Anti-TOM1L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
51017-1-AP-150UL
10156-150EA 635.16 CAD
10156-150
Anti-TOM1L1 Rabbit Polyclonal Antibody
Antibodies
The TOM1L1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TOM1L1. This antibody recognizes human antigen. The TOM1L1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300 ; IF 1 : 25

Type: Primary
Antigen: TOM1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR