Anti-ZNF323 Rabbit Polyclonal Antibody

Supplier: Proteintech
25593-1-AP-150UL
10155-712EA 635.16 CAD
10155-712
Anti-ZNF323 Rabbit Polyclonal Antibody
Antibodies
The ZNF323 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZNF323. This antibody recognizes human, mouse antigen. The ZNF323 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IHC 1 : 50

Type: Primary
Antigen: ZNF323
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR