Anti-ACTA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
55135-1-AP-150UL
10155-850EA 635.16 CAD
10155-850
Anti-ACTA1 Rabbit Polyclonal Antibody
Antibodies
The smooth muscle actin specific antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human smooth muscle actin specific. This antibody recognizes human,mouse,rat antigen. The smooth muscle actin specific antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 1000 ; IHC 1 : 50

Type: Primary
Antigen: ACTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR