Anti-ZNF645 Rabbit Polyclonal Antibody

Supplier: Proteintech
25600-1-AP-150UL
10155-718EA 635.16 CAD
10155-718
Anti-ZNF645 Rabbit Polyclonal Antibody
Antibodies
The ZNF645 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZNF645. This antibody recognizes human antigen. The ZNF645 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 3000

Type: Primary
Antigen: ZNF645
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR