Anti-PRND Rabbit Polyclonal Antibody

Supplier: Proteintech
51026-2-AP
10155-794EA 635.16 CAD
10155-794
Anti-PRND Rabbit Polyclonal Antibody
Antibodies
The Prnd antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of mouse Prnd. This antibody recognizes human, mouse antigen. The Prnd antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IHC 1 : 50 ; IP 1 : 300

Type: Primary
Antigen: PRND
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR