Anti-MUPCDH Rabbit Polyclonal Antibody

Supplier: Proteintech
25619-1-AP-150UL
10155-730EA 818.44 CAD
10155-730
Anti-MUPCDH Rabbit Polyclonal Antibody
Antibodies
The MUPCDH antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MUPCDH. This antibody recognizes human antigen. The MUPCDH antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600

Type: Primary
Antigen: MUPCDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR