Anti-STAT5A Rabbit Polyclonal Antibody

Supplier: Proteintech
51074-2-AP
10155-834EA 635.16 CAD
10155-834
Anti-STAT5A Rabbit Polyclonal Antibody
Antibodies
The STAT5A antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human STAT5A. This antibody recognizes human, mouse antigen. The STAT5A antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 50 ; IP 1 : 600

Type: Primary
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR