Anti-MARCKS Rabbit Polyclonal Antibody

Supplier: Proteintech
51087-1-AP
10156-338EA 635.16 CAD
10156-338
Anti-MARCKS Rabbit Polyclonal Antibody
Antibodies
The MARCKS antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human MARCKS. This antibody recognizes human, mouse antigen. The MARCKS antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300

Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR