Anti-HYAL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
51148-1-AP
10155-838EA 635.16 CAD
10155-838
Anti-HYAL2 Rabbit Polyclonal Antibody
Antibodies
The HYAL2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HYAL2. This antibody recognizes human, mouse antigen. The HYAL2 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: HYAL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR