Anti-STIM1 Mouse Monoclonal Antibody [clone: 1D3A3]

Supplier: Proteintech
66189-1-IG-150UL
10155-944EA 635.16 CAD
10155-944
Anti-STIM1 Mouse Monoclonal Antibody [clone: 1D3A3]
Antibodies
The STIM1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human STIM1. This antibody recognizes rat, human, mouse antigen. The STIM1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: STIM1
Clonality: Monoclonal
Clone: 1D3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR