Anti-Insulin Mouse Monoclonal Antibody [clone: 4B6A7]

Supplier: Proteintech
66198-1-IG
10155-962EA 818.44 CAD
10155-962
Anti-Insulin Mouse Monoclonal Antibody [clone: 4B6A7]
Antibodies
The Insulin antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human Insulin. This antibody recognizes human, mouse antigen. The Insulin antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 800 ; IHC 1 : 50

Type: Primary
Antigen: Insulin
Clonality: Monoclonal
Clone: 4B6A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR