Anti-TTC32 Rabbit Polyclonal Antibody

Supplier: Proteintech
25422-1-AP-150UL
10155-614EA 635.16 CAD
10155-614
Anti-TTC32 Rabbit Polyclonal Antibody
Antibodies
The TTC32 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TTC32. This antibody recognizes human, mouse antigen. The TTC32 antibody has been validated for the following applications: ELISA, WB, IHC, IP, IF analysis.

IHC 1 : 50 ; WB 1 : 3000

Type: Primary
Antigen: TTC32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR