Anti-RRP36 Rabbit Polyclonal Antibody

Supplier: Proteintech
25440-1-AP-150UL
10155-626EA 635.16 CAD
10155-626
Anti-RRP36 Rabbit Polyclonal Antibody
Antibodies
The RRP36 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RRP36. This antibody recognizes human antigen. The RRP36 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300

Type: Primary
Antigen: RRP36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR