Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Plastic Beads

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/56/18989656.jpg

Colored plastic beads can be a useful addition to the classroom in creating visual demonstrations of various procedures or internal biological systems, such as interactions between proteins and the proliferation of disease or a virus as it spreads through the body. They come in a variety of sorted colors, with precisely cut holes and uniform sizing. Plastic beads are economical, inert, and durable under conditions of constant, heavy use.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"5"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR
Description:   BEADS 8MM PKG/100 APPROX. NAVY
Supplier:  Thermo Scientific
Description:   These Iodination Beads are polystyrene beads that are coated with an oxidizing reagent, which provides efficient activation of 125I for pro...
SDS
Catalog Number: (470093-346)
Supplier:  DARICE
Description:   Pony Beads, Asst Colors.
Discontinued
Catalog Number: (470089-446)
Supplier:  PRECISION MOULDED POLYMERS LTD
Description:   Sorted colors. Plastic beads have 3.5mm hole and 13mm diameter. Set of 216.
Discontinued
Catalog Number: (470150-464)
Supplier:  American Scientific
Description:   Actually Change Color When Exposed to Sunlight
Small Business Enterprise
Limited Availability
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 5  of 5