Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VersaTouch® 87-297 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell

Supplier: Ansell Healthcare
VersaTouch®
192971 192973 0029711080 192975 0029711070 192977 0029711090 0029711100
Strength, sensitivity, moderate chemical protection and grip come together in a natural rubber latex glove that provides a high degree of tensile strength.

  • Excellent tactile sensitivity
  • Good protection against chemical splashes
  • Improved grip on wet or dry objects

The cotton flock lining helps maintain excellent sensitivity

Use in the automotive and transportation, chemical, food handling and processing, janitorial services, petrochemical, sanitation, and general industries. Uses include canning and food processing; chemical processing, oil refining and petrochemicals; handling acids, alkalies, salts, ketones and alcohols; and janitorial and maintenance work.

Gloves provide snag, puncture and abrasion resistance. The embossed pebble design provides improved grip on wet or dry objects.

Packaging: Gloves are packaged 12 pairs in a bag; 144 in a carton.



 

ORDER

Expand All / Collapse All
Glove Size Length Thickness Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
10 30.5 cm (12") 20 mil Yellow 0029711100 CA32890-542 Case of 144 Retrieving
Pack of 12 Retrieving
7 30.5 cm (12") 20 mil Yellow 0029711070 CA32890-536 Case of 144 Retrieving
Pack of 12 Retrieving
8 30.5 cm (12") 20 mil Yellow 0029711080 CA32890-538 Case of 144 Retrieving
Pack of 12 Retrieving
9 30.5 cm (12") 20 mil Yellow 0029711090 CA32890-540 Case of 144 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Hand-specific
Latex/Latex-free Latex
Liner material Cotton
Finish Pebble Embossed
Cuff style Pinked
EN norm 2110

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty