Samco™ Bio-Tite™ Specimen Container, 60 ml/48 mm (2 oz.), Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
BioTite™
03 0006 03 0193 03 0195 03 0007 03 0181
89498-366CS 624.67 CAD
89498-366 89498-364 89498-380 95043-676 89498-382
Samco™ Bio-Tite™ Specimen Container, 60 ml/48 mm (2 oz.), Thermo Fisher Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ Wide-Mouth 60mL (2oz.) 48mm Specimen Containers

  • Plug seal cap ensures excellent leak-resiatant performance
  • Cap and base knurls ensure ease of handling
  • Available either assembled or in bulk packed format ideal for pre-fill applications
  • Supplied nonsterile or gamma irradiated for guaranteed sterility

These stackable, shatter-resistant containers are manufactured from virgin polypropylene and are ideal for the collection and transportation of both liquid and solid samples. Available in both sterile and nonsterile formats, with a range of cap colors and label types to suit most applications.

Certifications: FDA Class 1 Exempt Medical Device.

Ordering information: Ambient storage and shipping recommended for all containers.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR