Anti-IgG1 lambda Rat Monoclonal Antibody [clone: RG1L]

Supplier: Rockland Immunochemicals
012-001-331 012-001-331
CA012-001-331EA 386.56 CAD
CA012-001-331 RL012-001-331
Anti-IgG1 lambda Rat Monoclonal Antibody [clone: RG1L]
Antibodies
Rat IgG1 Lambda isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG1 lambda
Clonality: Monoclonal
Clone: RG1L
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR