Anti-IgG2a Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
012-002-041 012-002-041
CA012-002-041EA 232.24 CAD
CA012-002-041 RL012-002-041
Anti-IgG2a Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Rat IgG2a isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!

Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR