Anti-IgG1 Rat Monoclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
012-006-040 012-006-040
CA012-006-040EA 231.93 CAD
CA012-006-040 RL012-006-040
Anti-IgG1 Rat Monoclonal Antibody (Biotin)
Antibodies
Rat IgG1 isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR