Anti-MYL12A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-416
CA10036-740EA 932.37 CAD
CA10036-740
Anti-MYL12A Rabbit Polyclonal Antibody
Antibodies
Anti-Myosin pS19/pS20 phospho specific polyclonal antibody was tested by ELISA, immunochemistry and immunoblotting.   Immunoblotting was used to show reactivity with unstimulated and stimulated cardiac myocytes.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 2.5 µg/ml; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MYL12A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR