Anti-TET2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-N93 100-401-N93
CA100-401-N93EA 779.7 CAD
RL100-401-N93 CA100-401-N93
Anti-TET2 Rabbit Polyclonal Antibody
Antibodies
Tet2 antibody has been tested for use by ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect band at ~230kDa and ~150kDa (isoforms).

Recommended Dilutions: ELISA: 1:50000; Western Blot: 1:5000;

Type: Primary
Antigen: TET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR