Anti-MMTV Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-P12
CA100-401-P12EA 779.7 CAD
CA100-401-P12
Anti-MMTV Rabbit Polyclonal Antibody
Antibodies
Anti-MMTV Antibody has been tested for use in western blotting. Specific conditions for reactivity should be optimized by the end user. Expect band approximately 26.7 kDa in size corresponding to MMTV capsid Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: MMTV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR