Anti-CODA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-094S 200-103-094S
CA10036-754EA 254.9 CAD
RL200-103-094S CA10036-754
Anti-CODA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Choline oxidase has been assayed against 1.0 µg of Choline Oxidase (Alcaligenes species) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:1000 - 1:2500; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: CODA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR