Anti-Protein A Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-106-077
CA200-106-077EA 835.65 CAD
CA200-106-077
Anti-Protein A Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Protein A Biotin Conjugated has been assayed against 1.0 µg of Protein A in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR