Anti-Protein A Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
200-109-077
CA200-109-077EA 835.65 CAD
CA200-109-077
Anti-Protein A Goat Polyclonal Antibody (Texas Red®)
Antibodies
Anti-Protein A Texa Red Conjugated Antibody is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR