Anti-GATA4 Mouse Monoclonal Antibody [clone: 10D8.B10.D4]

Supplier: Rockland Immunochemicals
200-301-P15S 200-301-P15S
CA200-301-P15SEA 213.16 CAD
CA200-301-P15S RL200-301-P15S
Anti-GATA4 Mouse Monoclonal Antibody [clone: 10D8.B10.D4]
Antibodies
Anti-GATA4 antibody is suitable for ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 44 kDa in size corresponding to GATA4 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: GATA4
Clonality: Monoclonal
Clone: 10D8.B10.D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR