Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 9G4.C3.F2]

Supplier: Rockland Immunochemicals
200-302-N61 200-302-N61
CA10036-898EA 677.57 CAD
CA10036-898 RL200-302-N61
Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 9G4.C3.F2]
Antibodies
Anti-CD8a is useful for immunochemistry and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/1x10e6 cells or 100µl of whole blood;
Do not freeze!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 9G4.C3.F2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR