Anti-CD3E Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: OKT3]

Supplier: Rockland Immunochemical
200-308-M62
CA10036-910EA 536.84 CAD
CA10036-910
Anti-CD3E Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: OKT3]
Antibodies
Anti-CD3 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/1x10^6 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR