Anti-MS4A1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: O323]

Supplier: Rockland Immunochemicals
200-308-N64 200-308-N64
CA10036-942EA 901.56 CAD
CA10036-942 RL200-308-N64
Anti-MS4A1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: O323]
Antibodies
Anti-CD20 is useful for Flow Cytometry, Immunoprecipitation, and immunochemistry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/1x10e6 cells or 100µl of whole blood;
Do not freeze! Light sensitive!

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: O323
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR