Anti-CD40 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5C3]

Supplier: Rockland Immunochemicals
200-308-N67 200-308-N67
CA200-308-N67EA 858.24 CAD
CA200-308-N67 RL200-308-N67
Anti-CD40 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5C3]
Antibodies
Anti-CD40 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR