Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HI30]

Supplier: Rockland Immunochemicals
200-308-N68 200-308-N68
CA200-308-N68EA 579.5 CAD
CA200-308-N68 RL200-308-N68
Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HI30]
Antibodies
Anti-CD45 is useful for Flow Cytometry, immunochemistry, and Western Blotting. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR